1-Indanone

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2,3-Dihydro-1H-inden-1-one

AS20084G100 AS20084G25
ACTIAS20084G100EA 0 GBP
ACTIAS20084G100 ACTIAS20084G25
1-Indanone
1-Indanone
Formula: C₉H₈O
MW: 132.16 g/mol
Boiling Pt: 243
Melting Pt: 37…40 °C
Density: 1.1028 g/cm³ (40 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003785
CAS Number: 83-33-0
EINECS: 201-470-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR