2,3-Dichloro-5-nitropyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS7041G1 AS7041G25 AS7041G5
ACTIAS7041G1EA 0 GBP
ACTIAS7041G1 ACTIAS7041G25 ACTIAS7041G5
2,3-Dichloro-5-nitropyridine
2,3-Dichloro-5-nitropyridine
Formula: C₅H₂Cl₂N₂O₂
MDL Number: MFCD03840432
CAS Number: 22353-40-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR