5-Bromo-2-nitropyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21163G100 AS21163G25
ACTIAS21163G100EA 0 GBP
ACTIAS21163G100 ACTIAS21163G25
5-Bromo-2-nitropyridine
5-Bromo-2-nitropyridine
Formula: C₅H₃BrN₂O₂
MDL Number: MFCD00160411
CAS Number: 39856-50-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR