Methyl-6-methylnicotinate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methyl 6-methylnicotinate

AS20588G100 AS20588G25
ACTIAS20588G100EA 0 GBP
ACTIAS20588G100 ACTIAS20588G25
Methyl-6-methylnicotinate
Methyl 6-methylnicotinate
Formula: C₈H₉NO₂
MW: 151.16 g/mol
MDL Number: MFCD00006340
CAS Number: 5470-70-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR