Methyl-5-methylnicotinate
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Methyl 5-methylnicotinate
AS21379G25
AS21379G5
ACTIAS21379G25EA
100
GBP
ACTIAS21379G25
ACTIAS21379G5
Methyl-5-methylnicotinate
Methyl 5-methylnicotinate
Formula:
C₈H₉NO₂ MW: 151.16 g/mol |
MDL Number:
MFCD01075739 CAS Number: 29681-45-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...