Methyl-2-aminonicotinate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methyl 2-aminonicotinate

AS22641G25 AS22641G5
ACTIAS22641G25EA 59 GBP
ACTIAS22641G25 ACTIAS22641G5
Methyl-2-aminonicotinate
Methyl 2-aminonicotinate
Formula: C₇H₈N₂O₂
MW: 152.15 g/mol
MDL Number: MFCD00128872
CAS Number: 14667-47-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR