2-Chloro-3-fluoroisonicotinic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Chloro-3-fluoropyridine-4-carboxylic acid

AS32439G1 AS32439G5
ACTIAS32439G1EA 12 GBP
ACTIAS32439G1 ACTIAS32439G5
2-Chloro-3-fluoroisonicotinic acid
2-Chloro-3-fluoroisonicotinic acid
Formula: C₆H₃ClFNO₂
MW: 175.55 g/mol
MDL Number: MFCD04038325
CAS Number: 628691-93-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR