2-Chloro-3-fluoro-4-pyridineboronic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Chloro-3-Fluoropyridine-4-Boronic Acid

AS22987G1 AS22987G25 AS22987G5
ACTIAS22987G1EA 24 GBP
ACTIAS22987G1 ACTIAS22987G25 ACTIAS22987G5
2-Chloro-3-fluoro-4-pyridineboronic acid
2-Chloro-3-fluoro-4-pyridineboronic acid
Formula: C₅H₄BClFNO₂
MW: 175.35 g/mol
MDL Number: MFCD04972403
CAS Number: 937595-71-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR