Methyl 2-amino-6-chloronicotinate
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Methyl 2-amino-6-chloronicotinate
AS12366G1
AS12366G5
ACTIAS12366G1EA
131
GBP
ACTIAS12366G1
ACTIAS12366G5
Methyl 2-amino-6-chloronicotinate
Methyl 2-amino-6-chloronicotinate
Formula:
C₇H₇ClN₂O₂ MW: 186.6 g/mol |
MDL Number:
MFCD17015967 CAS Number: 849805-25-0 |
Learn more
![](/m-uk.vwr.com/en_GB/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...