Methyl-6-chloronicotinate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS22422G100 AS22422G25
ACTIAS22422G100EA 0 GBP
ACTIAS22422G100 ACTIAS22422G25
Methyl-6-chloronicotinate
Methyl 6-chloronicotinate
Formula: C₇H₆ClNO₂
MDL Number: MFCD00023420
CAS Number: 73781-91-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR