Methyl-2,6-dichloronicotinate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methyl 2,6-dichloronicotinate

AS22391G1 AS22391G25 AS22391G5
ACTIAS22391G1EA 0 GBP
ACTIAS22391G1 ACTIAS22391G25 ACTIAS22391G5
Methyl-2,6-dichloronicotinate
Methyl 2,6-dichloronicotinate
Formula: C₇H₅Cl₂NO₂
MW: 206.03 g/mol
MDL Number: MFCD07369794
CAS Number: 65515-28-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR