Methyl-5-bromo-2-chloronicotinate
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS22713G1
AS22713G25
AS22713G5
ACTIAS22713G1EA
0
GBP
ACTIAS22713G1
ACTIAS22713G25
ACTIAS22713G5
Methyl-5-bromo-2-chloronicotinate
Methyl 5-bromo-2-chloronicotinate
Formula:
C₇H₅BrClNO₂ |
MDL Number:
MFCD03844848 CAS Number: 78686-79-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...