Iodosobenzene diacetate
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Iodobenzene diacetate, (Diacetoxyiodo)benzene
AS20580G100
AS20580G5
ACTIAS20580G100EA
0
GBP
ACTIAS20580G100
ACTIAS20580G5
Iodosobenzene diacetate
Iodosobenzene diacetate
Formula:
C₁₀H₁₁IO₄ MW: 322.1 g/mol |
MDL Number:
MFCD00008692 CAS Number: 3240-34-4 EINECS: 221-808-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...