(S)-(+)-2-Chloromandelic acid
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
(S)-2-(2-Chlorophenyl)-2-hydroxyacetic acid
AS22863G100
AS22863G25
ACTIAS22863G100EA
0
GBP
ACTIAS22863G100
ACTIAS22863G25
(S)-(+)-2-Chloromandelic acid
(S)-(+)-2-Chloromandelic acid
Formula:
C₈H₇ClO₃ MW: 186.59 g/mol |
MDL Number:
MFCD00798437 CAS Number: 52950-19-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...