(R)-(-)-2-Chloromandelic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (R)-2-(2-Chlorophenyl)-2-hydroxyacetic acid

AS22862G100 AS22862G25
ACTIAS22862G100EA 0 GBP
ACTIAS22862G100 ACTIAS22862G25
(R)-(-)-2-Chloromandelic acid
(R)-(-)-2-Chloromandelic acid
Formula: C₈H₇ClO₃
MW: 186.59 g/mol
MDL Number: MFCD00798429
CAS Number: 52950-18-2
EINECS: 444-040-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR