4-Methoxypicolinic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Methoxypyridine-2-carboxylic acid

AS1143G1 AS1143G25 AS1143G5
ACTIAS1143G1EA 0 GBP
ACTIAS1143G1 ACTIAS1143G25 ACTIAS1143G5
4-Methoxypicolinic acid
4-Methoxypicolinic acid
Formula: C₇H₇NO₃
MW: 153.14 g/mol
MDL Number: MFCD04115716
CAS Number: 29082-91-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR