(±)-β-Aminophenethyl alcohol
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
DL-2-Phenylglycinol, 2-Amino-2-phenylethanol, DL-Phenylglycinol
AS20221G100
AS20221G25
ACTIAS20221G100EA
0
GBP
ACTIAS20221G100
ACTIAS20221G25
(±)-β-Aminophenethyl alcohol
(±)-β-Aminophenethyl alcohol
Formula:
C₈H₁₁NO MW: 137.18 g/mol |
MDL Number:
MFCD00130145 CAS Number: 7568-92-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...