5-Fluoropicolinic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 5-Fluoro-2-pyridinecarboxylic acid, 5-Fluoro-2-picolinic acid, 5-Fluoropyridine-2-carboxylic acid

AS22828G1 AS22828G25 AS22828G5
ACTIAS22828G1EA 15 GBP
ACTIAS22828G1 ACTIAS22828G25 ACTIAS22828G5
5-Fluoropicolinic acid
5-Fluoropicolinic acid
Formula: C₆H₄FNO₂
MDL Number: MFCD04114196
CAS Number: 107504-08-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR