6-Bromopicolinic acid
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
6-Bromo-2-pyridinecarboxylic acid, 6-Bromopyridine-2-carboxylic acid
AS4019G100
AS4019G25
AS4019G5
ACTIAS4019G100EA
180
GBP
ACTIAS4019G100
ACTIAS4019G25
ACTIAS4019G5
6-Bromopicolinic acid
6-Bromopicolinic acid
Formula:
C₆H₄BrNO₂ MW: 202.01 g/mol |
MDL Number:
MFCD00093197 CAS Number: 21190-87-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...