4-Bromopicolinic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Bromopyridine-2-carboxylic acid

AS20516G25 AS20516G5
ACTIAS20516G25EA 82 GBP
ACTIAS20516G25 ACTIAS20516G5
4-Bromopicolinic acid
4-Bromopicolinic acid
Formula: C₆H₄BrNO₂
MW: 202.01 g/mol
MDL Number: MFCD03426665
CAS Number: 30766-03-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR