3-Bromopicolinic acid
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
3-Bromopyridine-2-carboxylic acid, 3-Bromo-2-pyridinecarboxylic acid
AS21430G1
AS21430G25
AS21430G5
ACTIAS21430G1EA
14
GBP
ACTIAS21430G1
ACTIAS21430G25
ACTIAS21430G5
3-Bromopicolinic acid
3-Bromopicolinic acid
Formula:
C₆H₄BrNO₂ MW: 202.01 g/mol |
MDL Number:
MFCD01320380 CAS Number: 30683-23-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...