(S)-(+)-β-Aminophenethyl alcohol

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (S)-(+)-2-Phenylglycinol, L-Phenylglycinol, (S)-2-Amino-2-phenylethanol

AS20176G100 AS20176G25
ACTIAS20176G100EA 0 GBP
ACTIAS20176G100 ACTIAS20176G25
(S)-(+)-β-Aminophenethyl alcohol
(S)-(+)-β-Aminophenethyl alcohol
Formula: C₈H₁₁NO
MW: 137.18 g/mol
MDL Number: MFCD00064404
CAS Number: 20989-17-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR