(R)-(-)-β-Aminophenethyl alcohol

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS20861G100 AS20861G25
ACTIAS20861G100EA 0 GBP
ACTIAS20861G100 ACTIAS20861G25
(R)-(-)-β-Aminophenethyl alcohol
(R)-(-)-β-Aminophenethyl alcohol
Formula: C₈H₁₁NO
MDL Number: MFCD00008062
CAS Number: 56613-80-0
EINECS: 260-287-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR