6-Bromoisatin

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21525G25 AS21525G5
ACTIAS21525G25EA 0 GBP
ACTIAS21525G25 ACTIAS21525G5
6-Bromoisatin
6-Bromoisatin
Formula: C₈H₄BrNO₂
MDL Number: MFCD01631138
CAS Number: 6326-79-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR