3-Amino-2-bromo-6-methylpyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS23647G1 AS23647G5
ACTIAS23647G1EA 0 GBP
ACTIAS23647G1 ACTIAS23647G5
3-Amino-2-bromo-6-methylpyridine
3-Amino-2-bromo-6-methylpyridine
Formula: C₆H₇BrN₂
MDL Number: MFCD03095078
CAS Number: 126325-53-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR