Tropine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21753G100 AS21753G25
ACTIAS21753G100EA 0 GBP
ACTIAS21753G100 ACTIAS21753G25
Tropine
Tropine
Formula: C₈H₁₅NO
MDL Number: MFCD00180667
CAS Number: 120-29-6
EINECS: 204-384-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR