Tropinone

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 8-Methyl-8-azabicyclo[3.2.1]octan-3-one

AS20052G100 AS20052G25
ACTIAS20052G100EA 0 GBP
ACTIAS20052G100 ACTIAS20052G25
Tropinone
Tropinone
Formula: C₈H₁₃NO
MW: 139.2 g/mol
MDL Number: MFCD00005549
CAS Number: 532-24-1
EINECS: 208-530-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR