Methyl-3-bromo-4-fluorobenzoate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methyl 3-bromo-4-fluorobenzoate

AS22637G25 AS22637G5
ACTIAS22637G25EA 0 GBP
ACTIAS22637G25 ACTIAS22637G5
Methyl-3-bromo-4-fluorobenzoate
Methyl 3-bromo-4-fluorobenzoate
Formula: C₈H₆BrFO₂
MDL Number: MFCD04972915
CAS Number: 82702-31-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR