4-Bromo-3-nitrobenzonitrile

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS22420G100 AS22420G25 AS22420G5
ACTIAS22420G100EA 330 GBP
ACTIAS22420G100 ACTIAS22420G25 ACTIAS22420G5
4-Bromo-3-nitrobenzonitrile
4-Bromo-3-nitrobenzonitrile
Formula: C₇H₃BrN₂O₂
MW: 227.02 g/mol
MDL Number: MFCD00982006
CAS Number: 89642-49-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR