Scandium(III) trifluoromethanesulphonate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Scandium(III) trifluoromethanesulphonate, Scandium triflate

AS20649G25 AS20649G5
ACTIAS20649G25EA 0 GBP
ACTIAS20649G25 ACTIAS20649G5
Scandium(III) trifluoromethanesulphonate
Scandium (III) trifluoromethanesulphonate
Formula: C₃F₉O₉S₃Sc
MDL Number: MFCD00192433
CAS Number: 144026-79-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR