1-Boc-4-hydroxypiperidine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS20187G100 AS20187G25
ACTIAS20187G100EA 0 GBP
ACTIAS20187G100 ACTIAS20187G25
1-Boc-4-hydroxypiperidine
1-Boc-4-hydroxypiperidine
Formula: C₁₀H₁₉NO₃
MDL Number: MFCD01075174
CAS Number: 109384-19-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR