1-Boc-3-hydroxypiperidine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: tert-butyl 3-hydroxypiperidine-1-carboxylate

AS20976G100 AS20976G25
ACTIAS20976G100EA 0 GBP
ACTIAS20976G100 ACTIAS20976G25
1-Boc-3-hydroxypiperidine
1-Boc-3-hydroxypiperidine
Formula: C₁₀H₁₉NO₃
MDL Number: MFCD02093938
CAS Number: 85275-45-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR