2,6-Dinitrobenzonitrile
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS20360G1
AS20360G25
AS20360G5
ACTIAS20360G1EA
0
GBP
ACTIAS20360G1
ACTIAS20360G25
ACTIAS20360G5
2,6-Dinitrobenzonitrile
2,6-Dinitrobenzonitrile
Formula:
C₇H₃N₃O₄ |
MDL Number:
MFCD00041246 CAS Number: 35213-00-4 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...