4-Bromo-3-hydroxybenzoic acid
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS23636G1
AS23636G5
ACTIAS23636G1EA
0
GBP
ACTIAS23636G1
ACTIAS23636G5
4-Bromo-3-hydroxybenzoic acid
4-Bromo-3-hydroxybenzoic acid
Formula:
C₇H₅BrO₃ |
MDL Number:
MFCD08056371 CAS Number: 14348-38-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...