4-Fluoro-3-nitrobenzonitrile

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS22419G100 AS22419G25 AS22419G5
ACTIAS22419G100EA 209 GBP
ACTIAS22419G100 ACTIAS22419G25 ACTIAS22419G5
4-Fluoro-3-nitrobenzonitrile
4-Fluoro-3-nitrobenzonitrile
Formula: C₇H₃FN₂O₂
MDL Number: MFCD01632197
CAS Number: 1009-35-4
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR