2,6-Difluoro-3-nitrobenzonitrile

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS24888G100 AS24888G25
ACTIAS24888G100EA 156 GBP
ACTIAS24888G100 ACTIAS24888G25
2,6-Difluoro-3-nitrobenzonitrile
2,6-Difluoro-3-nitrobenzonitrile
Formula: C₇H₂F₂N₂O₂
MDL Number: MFCD00042254
CAS Number: 143879-77-0
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR