2-Picolyl chloride hydrochloride

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-(Chloromethyl)pyridine HCl, 2-(Chloromethyl)pyridine hydrochloride

AS20073G100 AS20073G25
ACTIAS20073G100EA 0 GBP
ACTIAS20073G100 ACTIAS20073G25
2-Picolyl chloride hydrochloride
2-Picolyl chloride hydrochloride
Formula: C₆H₇Cl₂N
MDL Number: MFCD00012811
CAS Number: 6959-47-3
EINECS: 230-149-9
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR