Cyclobutanol
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS21221G1
AS21221G5
ACTIAS21221G1EA
0
GBP
ACTIAS21221G1
ACTIAS21221G5
Cyclobutanol
Cyclobutanol
Formula:
C₄H₈O MW: 72.11 g/mol Boiling Pt: 122 °C (1013 hPa) Density: 0.9226 g/cm³ (15 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00001318 CAS Number: 2919-23-5 EINECS: 220-858-1 UN: 1120 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...