Cyclobutanecarboxylic acid
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
AS20119G100
AS20119G25
ACTIAS20119G100EA
0
GBP
ACTIAS20119G100
ACTIAS20119G25
Cyclobutanecarboxylic acid
Cyclobutanecarboxylic acid
Formula:
C₅H₈O₂ MW: 100.12 g/mol Boiling Pt: 195 °C (1013 hPa) Melting Pt: –20 °C Density: 1.06 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00001323 CAS Number: 3721-95-7 EINECS: 223-072-7 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...