1,3-Cyclopentanedione

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Cyclopentane-1,3-dione

AS20785G25 AS20785G5
ACTIAS20785G25EA 73 GBP
ACTIAS20785G25 ACTIAS20785G5
1,3-Cyclopentanedione
1,3-Cyclopentanedione
Formula: C₅H₆O₂
MW: 98.1 g/mol
Melting Pt: 149…152 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001405
CAS Number: 3859-41-4
EINECS: 223-372-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR