Cyclobutanone

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS20518G25 AS20518G5
ACTIAS20518G25EA 0 GBP
ACTIAS20518G25 ACTIAS20518G5
Cyclobutanone
Cyclobutanone
Formula: C₄H₆O
Boiling Pt: 99 °C (1013 hPa)
Density: 0.93 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00001332
CAS Number: 1191-95-3
EINECS: 214-745-6
UN: 1224
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR