4-Methylquinoline
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
Lepidine
AS23284G100
AS23284G25
ACTIAS23284G100EA
149
GBP
ACTIAS23284G100
ACTIAS23284G25
4-Methylquinoline
4-Methylquinoline
Formula:
C₁₀H₉N MW: 143.19 g/mol Boiling Pt: 261 °C (1013 hPa) Melting Pt: 9 °C Density: 1.0995 g/cm³ (0 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00006784 CAS Number: 491-35-0 EINECS: 207-734-2 Merck Index: 13,05460 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...