5-Aminoquinoline

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Quinolin-5-amine, Quinolin-5-ylamine

AS7020G100 AS7020G25 AS7020G5
ACTIAS7020G100EA 275 GBP
ACTIAS7020G100 ACTIAS7020G25 ACTIAS7020G5
5-Aminoquinoline
5-Aminoquinoline
Formula: C₉H₈N₂
MW: 144.18 g/mol
Boiling Pt: 310 °C (1013 hPa)
Melting Pt: 107…109 °C
Storage Temperature: Ambient
MDL Number: MFCD00006797
CAS Number: 611-34-7
EINECS: 210-266-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR