5-Quinolinecarboxylic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

AS7096G1 AS7096G5
ACTIAS7096G1EA 30 GBP
ACTIAS7096G1 ACTIAS7096G5
5-Quinolinecarboxylic acid
5-Quinolinecarboxylic acid
Formula: C₁₀H₇NO₂
MW: 173.17 g/mol
MDL Number: MFCD00047608
CAS Number: 7250-53-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR