Quinaldic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

AS20072G100 AS20072G25
ACTIAS20072G100EA 0 GBP
ACTIAS20072G100 ACTIAS20072G25
Quinaldic acid
Quinaldic acid
Formula: C₁₀H₇NO₂
Melting Pt: 154…157 °C
Storage Temperature: Ambient
MDL Number: MFCD00006752
CAS Number: 93-10-7
EINECS: 202-218-3
Merck Index: 12,08227

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR