2-Phenylbenzoic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Biphenyl-2-carboxylic acid, 2-Biphenylcarboxylic acid, [1,1'-Biphenyl]-2-carboxylic acid

AS20985G100 AS20985G25
ACTIAS20985G100EA 0 GBP
ACTIAS20985G100 ACTIAS20985G25
2-Phenylbenzoic acid
2-Phenylbenzoic acid
Formula: C₁₃H₁₀O₂
MW: 198.22 g/mol
Boiling Pt: 344 °C (1013 hPa)
Melting Pt: 108…109 °C
Density: 1,458 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002463
CAS Number: 947-84-2
EINECS: 213-432-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR