2,3-Dimethylbenzoic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

AS21513G100 AS21513G25
ACTIAS21513G100EA 95 GBP
ACTIAS21513G100 ACTIAS21513G25
2,3-Dimethylbenzoic acid
2,3-Dimethylbenzoic acid
Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 282 °C (1013 hPa)
Melting Pt: 143…147 °C
Density: 1,283 g/cm³ (20 °C)
MDL Number: MFCD00002479
CAS Number: 603-79-2
EINECS: 210-058-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR