5-Methyl-2-thenoic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 5-Methylthiophene-2-carboxylic acid

AS20140G100 AS20140G25
ACTIAS20140G100EA 123 GBP
ACTIAS20140G100 ACTIAS20140G25
5-Methyl-2-thenoic acid
5-Methyl-2-thenoic acid
Formula: C₆H₆O₂S
MW: 142.18 g/mol
MDL Number: MFCD00005439
CAS Number: 1918-79-2
EINECS: 217-640-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR