4-Bromothiophene-2-carbaldehyde

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS20351G100 AS20351G25
ACTIAS20351G100EA 0 GBP
ACTIAS20351G100 ACTIAS20351G25
4-Bromothiophene-2-carbaldehyde
4-Bromothiophene-2-carbaldehyde
Formula: C₅H₃BrOS
MDL Number: MFCD00005431
CAS Number: 18791-75-8
EINECS: 242-577-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR