N-Acetyl-L-proline
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
N-Acetyl-L-Proline, Ac-Pro-OH
AS23050G100
AS23050G25
ACTIAS23050G100EA
0
GBP
ACTIAS23050G100
ACTIAS23050G25
N-Acetyl-L-proline
N-Acetyl-L-proline
Formula:
C₇H₁₁NO₃ MW: 157.169 g/mol |
MDL Number:
MFCD00020837 CAS Number: 68-95-1 EINECS: 276-259-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...